Assay Details
Target Gene Details
Entrez Gene ID: | 73181 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4 |
Gene Aliases: |
3110041H08Rik, AW107667, AW546455, Nfat3 |
Location: |
Chr.14:55824795-55833943 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfatc4 | NM_001168346.1 | 2 | 1251 | NP_001161818.1 |
| NM_023699.3 | 2 | 1251 | NP_076188.3 | |
| AK014164.1 | 1 | 490 | ||
| AK159078.1 | 2 | 1097 | BAE34797.1 | |
| BC028928.1 | 2 | 1038 | AAH28928.1 | |
| EU887656.1 | 2 | 1009 | ||
| EU887657.1 | 2 | 1009 |
More Information
Set Membership: |
|

Genomic Map