Assay Details
Target Gene Details
Entrez Gene ID: | 11827 |
Gene Name: | aquaporin 2 |
Gene Aliases: |
AQP-CD, WCH-CD, cph, jpk |
Location: |
Chr.15:99579056-99584545 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aqp2 | NM_009699.3 | 4 | 1299 | NP_033829.3 |
| AK002689.1 | 4 | 1300 | ||
| AK142567.1 | 4 | 1300 | BAE25108.1 | |
| BC019966.1 | 4 | 1283 | AAH19966.1 |
More Information
Set Membership: |
|

Genomic Map