Assay Details
Target Gene Details
Entrez Gene ID: | 223917 |
Gene Name: | keratin 79 |
Gene Aliases: |
BC031593 |
Location: |
Chr.15:101929332-101940324 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt79 | NM_146063.1 | 9 | 1900 | NP_666175.1 |
| AK076407.1 | 5 | 956 | ||
| BC019155.1 | 9 | 1839 | ||
| BC031593.1 | 9 | 1900 | AAH31593.1 |

Genomic Map