Assay Details
Target Gene Details
Entrez Gene ID: | 209200 |
Gene Name: | deltex 3-like (Drosophila) |
Gene Aliases: |
AU042200, BC023741 |
Location: |
Chr.16:35926515-35939027 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtx3l | NM_001013371.2 | 1 | 63 | NP_001013389.2 |
| BC137694.1 | ||||
| CN668148.1 | 1 | 63 |
Target Gene Details
Entrez Gene ID: | 80285 |
Gene Name: | poly (ADP-ribose) polymerase family, member 9 |
Gene Aliases: |
ARTD9, AW214463, BC003281, Bagl, Bal, PARP-9 |
Location: |
Chr.16:35938470-35972621 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp9 | NM_030253.3 | 1 | 496 | NP_084529.1 |
| AK037903.1 | BAC29897.1 | |||
| BC070466.1 | 1 | 496 | AAH70466.1 |
More Information
Set Membership: |
|

Genomic Map