Assay Details
Target Gene Details
Entrez Gene ID: | 13544 |
Gene Name: | dishevelled 3, dsh homolog (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.16:20516938-20532548 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dvl3 | NM_007889.2 | 1 | 142 | NP_031915.2 |
| AK052151.1 | 1 | 143 | ||
| U41285.1 | 1 | 224 | AAB01761.1 |
More Information
Set Membership: |
|

Genomic Map