Assay Details
Target Gene Details
Entrez Gene ID: | 12934 |
Gene Name: | dihydropyrimidinase-like 2 |
Gene Aliases: |
AI851130, Crmp2, DRP2, Musunc33, TOAD-64, Ulip2 |
Location: |
Chr.14:66802864-66910919 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpysl2 |
Target Gene Details
Entrez Gene ID: | 432870 |
Gene Name: | predicted gene 5464 |
Gene Aliases: |
EG432870 |
Location: |
Chr.14:66868850-66871005 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm5464 | NM_001034881.3 | 1 | 1792 | NP_001030053.1 |
| AK135012.1 | 1 | 1793 | BAE22384.1 |

Genomic Map