Assay Details
Target Gene Details
Entrez Gene ID: | 20220 |
Gene Name: | Sin3-associated polypeptide 18 |
Gene Aliases: |
C530046K05Rik, D11Ertd539e, EMegR4, Sinbp1 |
Location: |
Chr.14:57798189-57804980 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sap18 | NM_009119.3 | 4 | 2942 | NP_033145.2 |
| BM899282.1 | ||||
| Z97062.1 | 4 | 2877 | CAB09797.1 |
More Information
Set Membership: |
|

Genomic Map