Assay Details
Target Gene Details
Entrez Gene ID: | 66251 |
Gene Name: | ADP-ribosylation factor GTPase activating protein 3 |
Gene Aliases: |
0610009H19Rik, 1810004P07Rik, 1810035F16Rik, 9130416J18Rik, Arfgap1 |
Location: |
Chr.15:83299739-83350344 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgap3 | NM_025445.4 | 16 | 1826 | NP_079721.2 |
| AK002400.1 | 3 | 395 | ||
| AK007732.1 | 16 | 1746 | BAB25220.1 | |
| AK028990.1 | 16 | 1827 | BAC26229.1 | |
| AK075788.1 | 16 | 1812 | BAC35959.1 | |
| AK169107.1 | 16 | 1733 | BAE40889.1 | |
| AK194782.1 | 1 | 182 | ||
| BC004081.1 | 16 | 1762 | AAH04081.1 | |
| BC060369.1 | 16 | 1750 | AAH60369.1 |
More Information
Set Membership: |
|

Genomic Map