Assay Details
Target Gene Details
Entrez Gene ID: | 73167 |
Gene Name: | Rho GTPase activating protein 8 |
Gene Aliases: |
3110043J09Rik |
Location: |
Chr.15:84720052-84772857 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap8 | NM_001164627.1 | 13 | 1384 | NP_001158099.1 |
| NM_001164628.1 | 12 | 1376 | NP_001158100.1 | |
| NM_001205334.1 | 13 | 1404 | NP_001192263.1 | |
| NM_028455.4 | 12 | 1396 | NP_082731.2 | |
| AK014171.1 | 12 | 1347 | BAB29190.1 | |
| AK086816.1 | 12 | 1350 | BAC39750.1 | |
| AK088242.1 | 12 | 1352 | BAC40230.1 | |
| BC005563.1 | 11 | 1339 | AAH05563.1 | |
| BC010306.1 | 12 | 1366 | AAH10306.2 |
More Information
Set Membership: |
|

Genomic Map