Assay Details
Target Gene Details
Entrez Gene ID: | 78541 |
Gene Name: | ankyrin repeat and SOCS box-containing 8 |
Gene Aliases: |
4930539L19Rik, AI788835, C430011H06Rik |
Location: |
Chr.15:98134637-98165602 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb8 | NM_001170710.1 | 4 | 1870 | NP_001164181.1 |
| NM_001170711.1 | 4 | 1876 | NP_001164182.1 | |
| NM_030121.4 | 4 | 1866 | NP_084397.3 | |
| AK030168.1 | 4 | 1809 | BAC26819.1 | |
| BC025106.1 | 4 | 1790 | AAH25106.1 |

Genomic Map