Assay Details
Target Gene Details
Entrez Gene ID: | 70472 |
Gene Name: | ATPase family, AAA domain containing 2 |
Gene Aliases: |
2610509G12Rik |
Location: |
Chr.15:58094047-58135139 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atad2 | NM_027435.2 | 16 | 2157 | NP_081711.2 |
| AK029867.1 | 9 | 1304 | BAC26651.1 | |
| AK032877.1 | 16 | 2154 | ||
| AK037651.1 | 2 | 2679 | ||
| AK134319.1 | 3 | 533 | BAE22095.1 |
More Information
Set Membership: |
|

Genomic Map