Assay Details
Target Gene Details
Entrez Gene ID: | 70472 |
Gene Name: | ATPase family, AAA domain containing 2 |
Gene Aliases: |
2610509G12Rik |
Location: |
Chr.15:58094047-58135139 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atad2 | NM_027435.2 | 28 | 4424 | NP_081711.2 |
| BC032211.1 | 4 | 958 | AAH32211.1 | |
| BC071261.1 | 5 | 988 | AAH71261.1 |
More Information
Set Membership: |
|

Genomic Map