Assay Details
Target Gene Details
Entrez Gene ID: | 16680 |
Gene Name: | keratin 84 |
Gene Aliases: |
AV235125, HRb-1, Krt2-16, Krt2-3 |
Location: |
Chr.15:101516663-101532959 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt84 | NM_008474.2 | 1 | 527 | NP_032500.2 |
| AY028607.1 | 1 | 484 | AAK26233.1 | |
| BC114971.1 | 1 | 480 | AAI14972.1 | |
| CF173770.1 | 1 | 261 |

Genomic Map