Assay Details
Target Gene Details
Entrez Gene ID: | 16680 |
Gene Name: | keratin 84 |
Gene Aliases: |
AV235125, HRb-1, Krt2-16, Krt2-3 |
Location: |
Chr.15:101516663-101532959 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt84 | NM_008474.2 | 9 | 2402 | NP_032500.2 |
| AA738772.1 | 1 | 171 | ||
| AY028607.1 | 9 | 2359 | AAK26233.1 | |
| X65505.1 | 6 | 1422 | CAA46479.1 |

Genomic Map