Assay Details
Target Gene Details
Entrez Gene ID: | 268905 |
Gene Name: | keratin associated protein 13-1 |
Gene Aliases: |
AA589563, KAP13.1 |
Location: |
Chr.16:88728862-88729611 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap13-1 | NM_183189.1 | 1 | 626 | NP_899012.1 |
| BC027551.1 | 1 | 626 | AAH27551.1 |
More Information
Set Membership: |
|

Genomic Map