Assay Details
Target Gene Details
Entrez Gene ID: | 26560 |
Gene Name: | keratin associated protein 15 |
Gene Aliases: |
Krtap15-1, Pmg-2, Pmg2 |
Location: |
Chr.16:88829009-88829844 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap15 | NM_013713.1 | 1 | 139 | NP_038741.1 |
| AK132534.1 | 1 | 155 | BAE21225.1 | |
| BC116219.2 | 1 | 138 |
More Information
Set Membership: |
|

Genomic Map