Assay Details
Target Gene Details
Entrez Gene ID: | 170657 |
Gene Name: | keratin associated protein 19-1 |
Gene Aliases: |
Krtap16-2, Krtap16-9 |
Location: |
Chr.16:88868918-88869410 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap19-1 | NM_130876.3 | 1 | 245 | NP_570946.2 |
| AF345292.1 | 1 | 202 | AAK52890.1 |

Genomic Map