Assay Details
Target Gene Details
Entrez Gene ID: | 239932 |
Gene Name: | keratin associated protein 24-1 |
Gene Aliases: |
Gm312 |
Location: |
Chr.16:88610709-88612279 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap24-1 | NM_001163141.1 | 1 | 857 | NP_001156613.1 |
| AK037168.1 | 1 | 858 | BAC29730.1 |

Genomic Map