Assay Details
Target Gene Details
Entrez Gene ID: | 69533 |
Gene Name: | keratin associated protein 26-1 |
Gene Aliases: |
2310002B14Rik, AI507462 |
Location: |
Chr.16:88646824-88647796 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap26-1 | NM_027105.2 | 1 | 162 | NP_081381.1 |
| AK009086.1 | 1 | 164 | BAB26061.1 | |
| BC128283.1 | 1 | 154 | ||
| BC139410.1 | 1 | 157 | ||
| BC139411.1 | 1 | 157 |
More Information
Set Membership: |
|

Genomic Map