Assay Details
Target Gene Details
Entrez Gene ID: | 213484 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 18 |
Gene Aliases: |
BC036718 |
Location: |
Chr.14:70577847-70582571 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt18 | NM_153136.4 | 3 | 1556 | NP_694776.2 |
| AK089446.1 | 2 | 1344 | BAC40887.1 | |
| AK155086.1 | 3 | 1557 | BAE33037.1 | |
| AK157937.1 | 3 | 1557 | BAE34271.1 |
More Information
Set Membership: |
|

Genomic Map