Assay Details
Target Gene Details
Entrez Gene ID: | 239102 |
Gene Name: | zinc finger homeobox 2 |
Gene Aliases: |
ZFH-5 |
Location: |
Chr.14:55060265-55092433 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfhx2 | NM_001039198.1 | 2 | 1767 | NP_001034287.1 |
| AB190186.1 | 2 | 1767 | BAE78491.1 |
Target Gene Details
Entrez Gene ID: | 432855 |
Gene Name: | zinc finger homeobox 2, opposite strand |
Gene Aliases: |
Zfhx2as |
Location: |
Chr.14:55073091-55075874 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfhx2os | NR_004444.2 | 1 | 469 | |
| AK032589.1 | 1 | 469 |
More Information
Set Membership: |
|

Genomic Map