Assay Details
Target Gene Details
Entrez Gene ID: | 414066 |
Gene Name: | cDNA Sequence BC037032 |
Gene Aliases: |
- |
Location: |
Chr.15:4020111-4027406 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC037032 | NR_028266.1 | 1 | 842 | |
| AK038484.1 | 1 | 842 | ||
| AK142971.1 | 1 | 843 | ||
| AK161019.1 | 1 | 839 | ||
| AK169074.1 | 1 | 838 | ||
| AK169091.1 | 1 | 838 |
Target Gene Details
Entrez Gene ID: | 67041 |
Gene Name: | 3-oxoacid CoA transferase 1 |
Gene Aliases: |
2610008O03Rik, Oxct, Oxct2a, Scot-s |
Location: |
Chr.15:4026428-4155344 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxct1 | NM_024188.6 | 1 | 138 | NP_077150.1 |
| AK078086.1 | 1 | 139 |
More Information
Set Membership: |
|

Genomic Map