Assay Details
Target Gene Details
Entrez Gene ID: | 67041 |
Gene Name: | 3-oxoacid CoA transferase 1 |
Gene Aliases: |
2610008O03Rik, Oxct, Oxct2a, Scot-s |
Location: |
Chr.15:4026428-4155344 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxct1 | NM_024188.6 | 17 | 3217 | NP_077150.1 |
| AK010029.1 | 4 | 1600 | ||
| AK146158.1 | 17 | 2975 | BAE26941.1 | |
| AK151662.1 | 17 | 2956 | BAE30590.1 | |
| AK154327.1 | 1 | 1329 | ||
| BE952161.1 | 1 | 280 |
More Information
Set Membership: |
|

Genomic Map