Assay Details
Target Gene Details
Entrez Gene ID: | 16700 |
Gene Name: | keratin associated protein 6-1 |
Gene Aliases: |
HGTpII.1, HGTpII.2 |
Location: |
Chr.16:89031699-89032292 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap6-1 | NM_010672.3 | 1 | 492 | NP_034802.1 |
| BC107313.1 | ||||
| BC107314.1 | ||||
| CJ048388.1 | 1 | 532 | ||
| D86420.1 | 1 | 492 | BAA19681.1 |

Genomic Map