Assay Details
Target Gene Details
Entrez Gene ID: | 16701 |
Gene Name: | keratin associated protein 6-2 |
Gene Aliases: |
AI646275, HGTpII.4 |
Location: |
Chr.16:89419323-89420111 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap6-2 | NM_010673.2 | 1 | 662 | NP_034803.2 |
| AA763314.1 | 1 | 157 | ||
| D89902.1 | 2 | 602 | BAA20281.1 |

Genomic Map