Assay Details
Target Gene Details
Entrez Gene ID: | 71363 |
Gene Name: | keratin associated protein 7-1 |
Gene Aliases: |
5430433J05Rik |
Location: |
Chr.16:89507703-89508323 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap7-1 | NM_027771.1 | 1 | 317 | NP_082047.1 |
| AK017387.1 | 1 | 318 | BAB30724.1 | |
| CB320982.1 | 1 | 300 |

Genomic Map