Assay Details
Target Gene Details
Entrez Gene ID: | 16704 |
Gene Name: | keratin associated protein 19-5 |
Gene Aliases: |
Krtap8-2 |
Location: |
Chr.16:88895968-88896449 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krtap19-5 | NM_010676.2 | 1 | 239 | NP_034806.2 |
| AA717139.1 | 1 | 247 | ||
| D86422.1 | 1 | 250 | BAA19683.1 |

Genomic Map