Assay Details
Target Gene Details
Entrez Gene ID: | 219134 |
Gene Name: | shisa family member 2 |
Gene Aliases: |
9430059P22Rik, MAd2, Tmem46, mShisa, shisa |
Location: |
Chr.14:59625281-59631660 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shisa2 | NM_145463.5 | 1 | 571 | NP_663438.1 |
| AB280737.1 | 1 | 325 | ||
| AK034905.1 | 1 | 546 | BAC28876.1 | |
| AK087512.1 | 1 | 541 | BAC39906.1 | |
| BC024118.1 | AAH24118.1 | |||
| BC057640.1 | 1 | 135 | AAH57640.1 |
More Information
Set Membership: |
|

Genomic Map