Assay Details
Target Gene Details
Entrez Gene ID: | 53871 |
Gene Name: | polycystic kidney disease 2-like 2 |
Gene Aliases: |
TRPP5 |
Location: |
Chr.18:34409152-34442789 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkd2l2 | NM_001163004.1 | 4 | 515 | NP_001156476.1 |
| NM_016927.3 | 4 | 515 | NP_058623.2 | |
| AF182033.1 | 4 | 515 | AAF65621.1 | |
| AK133423.1 | 4 | 512 | BAE21649.1 | |
| AK142697.1 | 4 | 504 | BAE25165.1 | |
| BC138717.1 | 4 | 500 |
More Information
Set Membership: |
|

Genomic Map