Assay Details
Target Gene Details
Entrez Gene ID: | 114570 |
Gene Name: | cysteine-rich protein 3 |
Gene Aliases: |
TLP, TLP-A, TLP-B, Tlmp |
Location: |
Chr.17:46428195-46431780 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crip3 | NM_053250.2 | 8 | 879 | NP_444480.1 |
| NM_181664.2 | 8 | 820 | NP_858050.1 | |
| AF367970.1 | 8 | 820 | AAK60524.1 | |
| AF367971.1 | 8 | 879 | AAK60525.1 | |
| AK080272.1 | 8 | 879 | BAC37863.1 |
More Information
Set Membership: |
|

Genomic Map