Assay Details
Target Gene Details
Entrez Gene ID: | 14755 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class Q |
Gene Aliases: |
Gpi1, Gpi1h, Gpi1p, Gpih |
Location: |
Chr.17:25926420-25942877 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigq | NM_001291025.1 | 12 | 2180 | NP_001277954.1 |
| NM_011822.4 | 11 | 2000 | NP_035952.2 | |
| AB008895.1 | 11 | 1955 | BAA23615.1 | |
| AK155173.1 | 10 | 2181 | ||
| AK163680.1 | 11 | 1976 | BAE37455.1 | |
| AK169284.1 | 11 | 1983 | BAE41042.1 | |
| AK170431.1 | 12 | 2127 | BAE41791.1 | |
| AK189065.1 | 1 | 237 | ||
| AK210787.1 | 1 | 82 | ||
| AK212042.1 | 1 | 237 | ||
| AK213886.1 | 2 | 237 | ||
| AK215923.1 | 2 | 238 | ||
| BC003917.1 | 9 | 1090 | AAH03917.1 | |
| BC014287.1 | 11 | 1973 | AAH14287.1 |
More Information
Set Membership: |
|

Genomic Map