Assay Details
Target Gene Details
Entrez Gene ID: | 106894 |
Gene Name: | HMG box domain containing 3 |
Gene Aliases: |
2510002C16Rik, A630042L21Rik, AI413166, mKIAA0194 |
Location: |
Chr.18:61131277-61177050 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgxb3 | NM_134134.2 | 21 | 4452 | NP_598895.2 |
| NM_178277.1 | 20 | 4547 | NP_840061.1 | |
| AB045322.1 | 21 | 4453 | BAA97982.1 | |
| AK010887.1 | 1 | 58 | ||
| AK129085.1 | 17 | 3455 | BAC97895.1 | |
| AK132529.1 | 14 | 2931 | BAE21220.1 | |
| AK144993.1 | 18 | 3963 | ||
| BC025148.1 | 6 | 1415 | AAH25148.1 | |
| BC058089.1 | 18 | 3845 | AAH58089.1 | |
| BC060046.1 | 14 | 2877 | AAH60046.1 | |
| BC066029.1 | 15 | 3683 | AAH66029.1 | |
| BC079580.1 | 20 | 4495 | AAH79580.1 |

Genomic Map