Assay Details
Target Gene Details
Entrez Gene ID: | 78309 |
Gene Name: | cullin 9 |
Gene Aliases: |
1810035I07Rik, Cul-9, Parc, mKIAA0708 |
Location: |
Chr.17:46500609-46546388 on Build GRCm38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cul9 | NM_001081335.2 | 41 | 7759 | NP_001074804.2 |
| AK051474.1 | 18 | 2988 | ||
| AK122351.1 | 38 | 6749 | BAC65633.1 | |
| AK147675.1 | 39 | 8373 | ||
| AK162968.1 | 20 | 3221 | ||
| BP753198.1 | 1 | 104 |

Genomic Map