Assay Details
Target Gene Details
Entrez Gene ID: | 11692 |
Gene Name: | growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration) |
Gene Aliases: |
Alr, ERV1 |
Location: |
Chr.17:24693189-24696227 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfer | NM_023040.3 | 3 | 740 | NP_075527.2 |
| AA123449.1 | 1 | 95 | ||
| AB041561.1 | 3 | 657 | BAA95045.1 | |
| AF148688.1 | 3 | 520 | AAD36987.1 | |
| AK146579.1 | BAE27275.1 | |||
| BC023941.1 | 3 | 740 | AAH23941.1 | |
| BI689446.1 | 1 | 219 | ||
| CA321838.1 | 1 | 205 |
More Information
Set Membership: |
|

Genomic Map