Assay Details
Target Gene Details
Entrez Gene ID: | 27361 |
Gene Name: | methionine sulfoxide reductase B1 |
Gene Aliases: |
D17Wsu82e, SELX, SelR, Sepr, Sepx1 |
Location: |
Chr.17:24736642-24742778 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msrb1 | NM_013759.2 | 5 | 805 | NP_038787.1 |
| AF195142.1 | 5 | 780 | AAF13697.1 | |
| AK002339.1 | 5 | 765 | BAC55245.1 | |
| AK002652.1 | 5 | 764 | BAE43148.1 | |
| AK003112.1 | 4 | 1137 | BAC55248.1 | |
| AK003624.1 | 5 | 765 | BAC55249.1 | |
| AK007404.1 | 1 | 324 | ||
| BC090646.1 | 5 | 805 | ||
| BC141146.1 |
More Information
Set Membership: |
|

Genomic Map