Assay Details
Target Gene Details
Entrez Gene ID: | 353502 |
Gene Name: | host cell factor C1 regulator 1 (XPO1-dependent) |
Gene Aliases: |
HPIP |
Location: |
Chr.17:23673608-23675524 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcfc1r1 | NM_181821.1 | NP_861542.1 | ||
| AK013438.1 | BAB28855.1 |
Target Gene Details
Entrez Gene ID: | 386612 |
Gene Name: | THO complex 6 homolog (Drosophila) |
Gene Aliases: |
F830014G06Rik, Wdr58 |
Location: |
Chr.17:23668618-23673903 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thoc6 | NM_001008425.1 | 1 | 130 | NP_001008425.1 |
| AK089746.1 | 1 | 203 | ||
| BC085131.1 | 1 | 130 | AAH85131.1 | |
| BC089551.1 | 2 | 130 | AAH89551.1 |

Genomic Map