Assay Details
Target Gene Details
Entrez Gene ID: | 11555 |
Gene Name: | adrenergic receptor, beta 2 |
Gene Aliases: |
Adrb-2, Badm, Gpcr7 |
Location: |
Chr.18:62177713-62179981 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adrb2 | NM_007420.3 | 1 | 1100 | NP_031446.2 |
| AK080241.1 | 1 | 1098 | BAC37856.1 | |
| AK080276.1 | 1 | 1099 | ||
| BC030346.1 | 1 | 536 | AAH30346.1 | |
| BC032883.1 | 1 | 1078 | AAH32883.1 |
More Information
Set Membership: |
|

Genomic Map