Assay Details
Target Gene Details
Entrez Gene ID: | 15526 |
Gene Name: | heat shock protein 9 |
Gene Aliases: |
74kDa, Csa, Grp75, Hsc74, Hsp74, Hsp74a, Hspa9a, Mortalin, Mot-2, Mot2, Mthsp70, Pbp74 |
Location: |
Chr.18:34937414-34954351 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa9 | NM_010481.2 | 17 | 2561 | NP_034611.2 |
| AK004946.1 | 17 | 2523 | BAB23690.1 | |
| AK030299.1 | 17 | 2555 | ||
| AK145965.1 | 17 | 2555 | BAE26790.1 | |
| AK159791.1 | 17 | 2563 | BAE35373.1 | |
| AK161587.1 | 17 | 2564 | ||
| AK165958.1 | 17 | 2561 | BAE38486.1 | |
| AK167856.1 | 17 | 2554 | BAE39874.1 | |
| BC052727.1 | 17 | 2561 | AAH52727.1 | |
| BC057343.1 | 17 | 2561 | AAH57343.1 | |
| BP769296.1 | ||||
| D17556.1 | 17 | 2533 | BAA04493.1 |

Genomic Map