Assay Details
Target Gene Details
Entrez Gene ID: | 74002 |
Gene Name: | pleckstrin and Sec7 domain containing 2 |
Gene Aliases: |
6330404E20Rik, AW125584, EFA6C |
Location: |
Chr.18:35963881-36014715 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psd2 | NM_001289602.1 | 2 | 733 | NP_001276531.1 |
| NM_028707.4 | 2 | 730 | NP_082983.3 | |
| NR_110349.1 | 2 | 730 | ||
| AK034575.1 | 2 | 496 | ||
| AK045529.1 | 2 | 458 | BAC32407.1 | |
| AK053835.1 | 2 | 458 | ||
| AK149027.1 | 2 | 456 | ||
| BC062930.1 | 1 | 311 | AAH62930.1 | |
| BC065055.1 | 2 | 481 | AAH65055.1 | |
| BC066026.1 | 2 | 730 | AAH66026.1 | |
| BC066036.1 | 2 | 496 | AAH66036.1 |
More Information
Set Membership: |
|

Genomic Map