Assay Details
Target Gene Details
Entrez Gene ID: | 13078 |
Gene Name: | cytochrome P450, family 1, subfamily b, polypeptide 1 |
Gene Aliases: |
CP1B, P4501b1 |
Location: |
Chr.17:79696282-79715065 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp1b1 | NM_009994.1 | 1 | 175 | NP_034124.1 |
| AK137461.1 | 1 | 94 | BAE23362.1 | |
| AK139368.1 | 1 | 196 | BAE23978.1 | |
| BC050063.1 | 1 | 175 | AAH50063.1 | |
| U03283.1 | 1 | 175 | AAC52141.1 | |
| X78445.1 | CAA55205.1 |
More Information
Set Membership: |
|

Genomic Map