Assay Details
Target Gene Details
Entrez Gene ID: | 56050 |
Gene Name: | cytochrome P450, family 39, subfamily a, polypeptide 1 |
Gene Aliases: |
mCYP39A1 |
Location: |
Chr.17:43667372-43751431 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp39a1 | NM_001285947.1 | 1 | 236 | NP_001272876.1 |
| NM_001285948.1 | 1 | 236 | NP_001272877.1 | |
| NM_018887.4 | 1 | 236 | NP_061375.1 | |
| AF237981.1 | 1 | 85 | AAF63328.1 | |
| AK031733.1 | 1 | 184 | BAC27530.1 | |
| BC021418.1 | 1 | 107 | ||
| BC037663.1 | 1 | 85 | AAH37663.1 | |
| BE648427.1 | 2 | 287 |
More Information
Set Membership: |
|

Genomic Map