Assay Details
Target Gene Details
Entrez Gene ID: | 106522 |
Gene Name: | protein kinase domain containing, cytoplasmic |
Gene Aliases: |
AI115348, AW548124, Adtk1, ESTM17, MAd1, Sgk493, Vlk, X83346 |
Location: |
Chr.17:83215283-83225069 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkdcc | NM_134117.2 | 3 | 1171 | NP_598878.2 |
| AB381893.1 | 3 | 1163 | ||
| AB436780.1 | 3 | 1152 | ||
| BC022157.1 | 2 | 318 | AAH22157.1 | |
| BC086639.1 | 4 | 1009 | AAH86639.1 |

Genomic Map