Assay Details
Target Gene Details
Entrez Gene ID: | 225027 |
Gene Name: | serine/arginine-rich splicing factor 7 |
Gene Aliases: |
35kDa, 9430065L19Rik, 9G8, NX-96, Sfrs7 |
Location: |
Chr.17:80200080-80207318 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf7 | NM_001195485.1 | 8 | 2032 | NP_001182414.1 |
| NM_001195486.1 | 8 | 2008 | NP_001182415.1 | |
| NM_001195487.1 | 7 | 1996 | NP_001182416.1 | |
| NM_146083.2 | 8 | 2041 | NP_666195.1 | |
| NR_036615.1 | 9 | 2492 | ||
| AK045884.1 | 8 | 2203 | BAC32521.1 | |
| AK161652.1 | 8 | 2040 | ||
| BC025529.1 | 8 | 2032 | AAH25529.1 | |
| BX519093.1 |
More Information
Set Membership: |
|

Genomic Map