Assay Details
Target Gene Details
Entrez Gene ID: | 57262 |
Gene Name: | resistin like alpha |
Gene Aliases: |
1810019L16Rik, Fizz-1, Fizz1, HIMF, RELM-alpha, RELMa, RELMalpha, Xcp2 |
Location: |
Chr.16:48842552-48844461 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Retnla | NM_020509.3 | 4 | 547 | NP_065255.2 |
| AF205951.1 | 3 | 480 | AAG02143.1 | |
| AF290872.1 | 4 | 585 | AAK83104.1 | |
| AF316397.2 | 4 | 489 | AAG41427.1 | |
| AF323082.1 | 4 | 557 | AAG59825.1 | |
| AK007553.1 | 4 | 492 | BAB25104.1 | |
| BC029248.1 | 3 | 487 | AAH29248.2 |

Genomic Map