Assay Details
Target Gene Details
Entrez Gene ID: | 68617 |
Gene Name: | microtubule crosslinking factor 1 |
Gene Aliases: |
1110012J17Rik, Ccdc165, D130071O13, Kiaa0802, Soga2, mKIAA0802, t8219b25 |
Location: |
Chr.17:66336980-66449808 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtcl1 | NM_001114098.1 | 15 | 6693 | NP_001107570.1 |
| NM_172963.4 | 14 | 6634 | NP_766551.3 | |
| AK003637.1 | 1 | 536 | ||
| AK122373.1 | 4 | 2204 | BAC65655.1 | |
| AK147205.1 | 14 | 6636 | BAE27762.1 | |
| AK147631.1 | 15 | 6685 | ||
| AK147691.1 | 15 | 6685 | ||
| BC060225.1 | 12 | 4882 | AAH60225.1 | |
| BP760436.1 |

Genomic Map