Assay Details
Target Gene Details
Entrez Gene ID: | 14990 |
Gene Name: | histocompatibility 2, M region locus 2 |
Gene Aliases: |
H-2M2, Thy19.4 |
Location: |
Chr.17:37480851-37483624 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-M2 | NM_008204.2 | 3 | 585 | NP_032230.1 |
| AY302212.1 | 3 | 585 | AAQ81303.1 | |
| AY302213.1 | 3 | 585 | AAQ81304.1 | |
| AY302214.1 | 3 | 585 | AAQ81305.1 | |
| AY302215.1 | 3 | 585 | AAQ81306.1 | |
| AY302216.1 | 3 | 585 | AAQ81307.1 | |
| AY302217.1 | 3 | 585 | AAQ81308.1 | |
| BC150676.1 | 3 | 608 |
More Information
Set Membership: |
|

Genomic Map