Assay Details
Target Gene Details
Entrez Gene ID: | 20473 |
Gene Name: | sine oculis-related homeobox 3 |
Gene Aliases: |
E130112M24Rik, Six3a, Six3alpha, Six3b, Six3beta |
Location: |
Chr.17:85620834-85626191 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Six3 | NM_011381.4 | 2 | 1283 | NP_035511.2 |
| BC098096.1 | 2 | 903 | AAH98096.1 | |
| CD352849.1 | 2 | 341 | ||
| D83145.1 | 2 | 869 | BAA11823.1 |
More Information
Set Membership: |
|

Genomic Map