Assay Details
Target Gene Details
Entrez Gene ID: | 328789 |
Gene Name: | lipoma HMGIC fusion partner-like 5 |
Gene Aliases: |
9330179O15Rik, Tmhs, hscy |
Location: |
Chr.17:28575371-28583593 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lhfpl5 | NM_026571.2 | 4 | 1012 | NP_080847.2 |
| AK020389.1 | 4 | 1018 | ||
| AK020670.1 | 4 | 1000 | ||
| BC099483.1 | 4 | 910 | AAH99483.1 |

Genomic Map