Assay Details
Target Gene Details
Entrez Gene ID: | 353502 |
Gene Name: | host cell factor C1 regulator 1 (XPO1-dependent) |
Gene Aliases: |
HPIP |
Location: |
Chr.17:23673608-23675524 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcfc1r1 | NM_181821.1 | NP_861542.1 | ||
| AK013438.1 | BAB28855.1 |
Target Gene Details
Entrez Gene ID: | 27279 |
Gene Name: | tumor necrosis factor receptor superfamily, member 12a |
Gene Aliases: |
AI255180, C87282, Fn14, HPIP, TWEAK-R, TweakR |
Location: |
Chr.17:23675445-23677449 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf12a | NM_001161746.1 | 3 | 763 | NP_001155218.1 |
| NM_013749.2 | 4 | 868 | NP_038777.2 | |
| AF156164.1 | 4 | 863 | AAF07882.1 | |
| AI853177.1 | 1 | 113 | ||
| AK005382.1 | 4 | 865 | BAB23989.1 | |
| AK005530.1 | 4 | 864 | BAB24101.1 | |
| AK160136.1 | 4 | 869 | BAE35650.1 | |
| BC025860.1 | 4 | 852 | AAH25860.1 |
More Information
Set Membership: |
|

Genomic Map