Assay Details
Target Gene Details
Entrez Gene ID: | 27279 |
Gene Name: | tumor necrosis factor receptor superfamily, member 12a |
Gene Aliases: |
AI255180, C87282, Fn14, HPIP, TWEAK-R, TweakR |
Location: |
Chr.17:23675445-23677449 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf12a | NM_001161746.1 | 3 | 370 | NP_001155218.1 |
| NM_013749.2 | 4 | 475 | NP_038777.2 | |
| AF156164.1 | 4 | 470 | AAF07882.1 | |
| AK005382.1 | 4 | 472 | BAB23989.1 | |
| AK005530.1 | 4 | 471 | BAB24101.1 | |
| AK160136.1 | 4 | 476 | BAE35650.1 | |
| BC025860.1 | 4 | 459 | AAH25860.1 | |
| DV655788.1 | 3 | 350 |
More Information
Set Membership: |
|

Genomic Map